OriGene
Manufacturer Code:TA190077
Catalog # 50-167-1211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F7 |
Immunogen | Synthetic peptide corresponding to the conserved region of human neurofilament heavy polypeptide (NEFH) medium polypeptide (NEFM) and light polypeptide (NF68 NEFL) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 160 kDa neurofilament protein; 200 kDa neurofilament protein; 68 kDa neurofilament protein; light molecular weight neurofilament protein; Neurofilament; Neurofilament 3; Neurofilament heavy polypeptide; Neurofilament light polypeptide; Neurofilament medium polypeptide; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet H protein; Neurofilament triplet L protein; Neurofilament triplet M protein; neurofilament, heavy polypeptide 200kDa; neurofilament, light polypeptide 68kDa; neurofilament, medium polypeptide 150kDa; neurofilament-3 (150 kD medium); NF-H; NF-L; NF-M; protein phosphatase 1, regulatory subunit 110
Gene Aliases: CMT1F; CMT2CC; CMT2E; KIAA0845; NEF3; NEFH; NEFL; NEFM; NF-L; NF-M; NF68; NFH; NFL; NFM; PPP1R110
UniProt ID: (Human) P12036, (Human) P07196, (Human) P07197
Entrez Gene ID: (Human) 4744, (Human) 4747, (Human) 4741
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.