Invitrogen
Manufacturer Code:PA540540
Catalog # PIPA540540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NEUROD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atonal homolog 2; bHLHa2; brain my051 protein; Class A basic helix-loop-helix protein 2; Helix-loop-helix protein mATH-2; mATH2; NeuroD6; neurogenic differentiation 6; Neurogenic differentiation factor 6; Neuronal differentiation 6 Nex1 Atoh2 MATH2 NEX1M Math-2 bHLHa2; Protein atonal homolog 2; Protein NEX-1
Gene Aliases: ATH2; ATOH2; BHLHA2; Math-2; MATH2; My051; NEUROD6; Nex; Nex1; NEX1M; RGD1562793
UniProt ID: (Bovine) Q08DI0, (Human) Q96NK8, (Mouse) P48986
Entrez Gene ID: (Dog) 482387, (Bovine) 540464, (Human) 63974, (Pig) 110257619, (Horse) 100055160, (Mouse) 11922, (Rat) 500137
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.