OriGene
Manufacturer Code:CF810658
Catalog # 50-167-0231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B7 |
Immunogen | Full length human recombit protein of human NEUROD4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATH-3; ATH-3 ATH3 Atoh3 MATH-3 MATH3 bHLHa4; bHLHa4; Class A basic helix-loop-helix protein 4; NeuroD4; neurogenic differentiation 4; Neurogenic differentiation factor 4; Protein atonal homolog 3
Gene Aliases: ATH-3; ATH3; ATOH3; BHLHA4; MATH-3; MATH3; NEUROD4
UniProt ID: (Human) Q9HD90
Entrez Gene ID: (Human) 58158
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.