Proteintech
Manufacturer Code:146101AP150UL
Catalog # 50-172-9582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NEUROD4 Fusion Protein Ag6183 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATH 3 ATH3 Atoh3 bHLHa4 MATH 3 NEUROD4 neurogenic differentiation 4 Protein atonal homolog 3; ATH-3; atonal homolog 3; bHLHa4; Class A basic helix-loop-helix protein 4; Helix-loop-helix protein mATH-3; mATH3; NeuroD4; neurogenic differentiation 4; Neurogenic differentiation factor 4; Protein atonal homolog 3
Gene Aliases: AI846749; ATH-3; ATH3; ATOH3; BHLHA4; MATH-3; MATH3; NEUROD4
UniProt ID: (Human) Q9HD90, (Mouse) O09105
Entrez Gene ID: (Human) 58158, (Mouse) 11923, (Rat) 288821
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.