Invitrogen
Manufacturer Code:PA571797
Catalog # PIPA571797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 234-262 amino acids from the C-terminal region of human NEURL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase NEURL3; E3 ubiquitin-protein ligase NEURL3 632- Lung-inducible neuralized-related C3CH4 RING domain protein Neuralized-like protein 3 NEURL3 LINCR; Lung-inducible neuralized-related C3CH4 RING domain protein; neuralized homolog 3 pseudogene; Neuralized-like protein 3; RING-type E3 ubiquitin transferase NEURL3
Gene Aliases: LINCR; NEURL3
UniProt ID: (Human) Q96EH8
Entrez Gene ID: (Human) 93082
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.