Invitrogen
Manufacturer Code:PA5101706
Catalog # PIPA5101706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NEURL2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf163 Neur2 neuralized E3 ubiquitin protein ligase 2 neuralized homolog 2 neuralized-like 2 neuralized-like 2 (Drosophila) neuralized-like protein 2 NEURL2 OZZ Ozz-E3; neuralized homolog 2; neuralized-like 2; Neuralized-like protein 2
Gene Aliases: C20orf163; Neur2; NEURL2; OZZ; OZZ-E3
UniProt ID: (Human) Q9BR09
Entrez Gene ID: (Human) 140825, (Mouse) 415115, (Rat) 311633
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.