Novus Biologicals
Manufacturer Code:NBP287899
Catalog # NB035521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human NEU3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.1.18 FLJ12388 Ganglioside sialidase Membrane sialidase N-acetyl-alpha-neuraminidase 3 neuraminidase 3 SIAL3 sialidase 3 (membrane sialidase) sialidase-3; ganglioside sialidase; Ganglioside sialidasedis; Membrane sialidase; N-acetyl-alpha-neuraminidase 3; neuraminidase 3 (membrane sialidase); neuraminidase 3, membrane sialidase; sialidase 3 (membrane sialidase); Sialidase-3
Gene Aliases: NEU3; SIAL3
UniProt ID: (Human) Q9UQ49
Entrez Gene ID: (Human) 10825
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.