Novus Biologicals
Manufacturer Code:NBP272944APC
Catalog # NB272944APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F4 |
Immunogen | Full length recombinant protein of human NEU2 (NP_005374) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cytosolic sialidase; Cytosolic sialidase EC 3.2.1.18 N-acetyl-alpha-neuraminidase 2 neuraminidase 2 SIAL2MGC129579 sialidase 2 (cytosolic sialidase) sialidase-2; N-acetyl-alpha-neuraminidase 2; neuraminidase 2 (cytosolic sialidase); sialidase 2 (cytosolic sialidase); Sialidase-2
Gene Aliases: NEU2; SIAL2
UniProt ID: (Human) Q9Y3R4
Entrez Gene ID: (Human) 4759
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.