Invitrogen
Manufacturer Code:PA535114
Catalog # PIPA535114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 23-50 amino acids from the N-terminal region of human NEU2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; NEU2 Sialidase-2 Cytosolic sialidase N-acetyl-alpha-neuraminidase 2; neuraminidase 2 (cytosolic sialidase); sialidase 2 (cytosolic sialidase); Sialidase-2
Gene Aliases: NEU2; SIAL2
UniProt ID: (Human) Q9Y3R4
Entrez Gene ID: (Human) 4759
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.