Invitrogen
Manufacturer Code:PA542552
Catalog # PIPA542552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NEU1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylneuraminyl hydrolase; exo-alpha-sialidase; G9 sialidase; HLA-B-associated transcript 7; Lysosomal sialidase; N-acetyl-alpha-neuraminidase 1; sialidase 1; sialidase 1 (lysosomal sialidase); Sialidase 1 (lysosomal sialidase) NEU SIAL1 NANH; Sialidase-1
Gene Aliases: AA407268; AA407316; Aglp; Apl; Bat-7; Bat7; G9; Map-2; NANH; NEU; Neu-1; NEU1; SIAL1
UniProt ID: (Human) Q99519, (Bovine) A6BMK7, (Mouse) O35657
Entrez Gene ID: (Dog) 481717, (Human) 4758, (Rabbit) 100342130, (Horse) 100059083, (Bovine) 505554, (Mouse) 18010, (Guinea pig) 100728899, (Rat) 24591
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.