Invitrogen
Manufacturer Code:PA5111787
Catalog # PIPA5111787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 189-202 of rat NETO2 (Extracellular N-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5530601C23Rik AI854825 Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2 Btcl2 FLJ10430 FLJ14724 FLJ90456 NEOT2 NEOT2FLJ14724 Neto2 Neuropilin and tolloid-like protein 2 UNQ1926/PRO4401; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; neuropilin (NRP) and tolloid (TLL)-like 2; neuropilin and tolloid like-2; Neuropilin and tolloid-like protein 2
Gene Aliases: 5530601C23Rik; AI854825; BTCL2; NEOT2; NETO2; UNQ1926/PRO4401
UniProt ID: (Human) Q8NC67, (Rat) C6K2K4, (Mouse) Q8BNJ6
Entrez Gene ID: (Human) 81831, (Rat) 307757, (Mouse) 74513
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.