Novus Biologicals
Manufacturer Code:NBP162591
Catalog # NBP162591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NETO2(neuropilin (NRP) and tolloid (TLL)-like 2) The peptide sequence was selected from the N terminal of NETO2. Peptide sequence ELSGADGIVRSSQVEQEEKTKPGQAVDCIWTIKATPKAKIYLRFLDYQME. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 2; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 2 BTCL2 FLJ10430 FLJ90456 NEOT2FLJ14724 neuropilin (NRP) and tolloid (TLL)-like 2 neuropilin and tolloid-like protein 2; neuropilin (NRP) and tolloid (TLL)-like 2; Neuropilin and tolloid-like protein 2
Gene Aliases: BTCL2; NEOT2; NETO2; UNQ1926/PRO4401
UniProt ID: (Human) Q8NC67
Entrez Gene ID: (Human) 81831
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.