R&D Systems
Manufacturer Code:MAB4337SP
Catalog # MAB4337SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 532509 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human NETO1 isoform 3 Thr23-Val345 Accession # Q8TDF5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCTL1 Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1 BTCL1 BTCL1BCTL1neuropilin and tolloid-like protein 1 FLJ41325 neuropilin (NRP) and tolloid (TLL)-like 1; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; neuropilin (NRP) and tolloid (TLL)-like 1; Neuropilin and tolloid-like protein 1
Gene Aliases: BCTL1; BTCL1; NETO1
UniProt ID: (Human) Q8TDF5
Entrez Gene ID: (Human) 81832
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.