Novus Biologicals
Manufacturer Code:NBP299822IR
Catalog # NB089636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant NETO1 (NP_620416.1 Met 1-Thr 344). |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1 BTCL1BCTL1neuropilin and tolloid-like protein 1 FLJ41325 neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; Neuropilin and tolloid-like protein 1
Gene Aliases: BCTL1; BTCL1; NETO1
UniProt ID: (Human) Q8TDF5
Entrez Gene ID: (Human) 81832
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.