Novus Biologicals
Manufacturer Code:NB1001328
Catalog # NB1001328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DKALSGGKRKETLV corresponding to C-Terminus according to NP_001040625.1 NP_005854.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGEF8neuroepithelial cell-transforming gene 1 protein guanine nucleotide regulatory protein (oncogene) NET1A neuroepithelial cell transforming 1 neuroepithelial cell transforming gene 1 neuroepithelioma transforming gene 1 p65 Net1 proto-oncogene protein Proto-oncogene p65 Net1 Rho guanine nucleotide exchange factor (GEF) 8 Rho guanine nucleotide exchange factor 8 small GTP-binding protein regulator; guanine nucleotide regulatory protein (oncogene); Neuroepithelial cell-transforming gene 1 protein; neuroepithelioma transforming gene 1; p65 Net1 proto-oncogene protein; Proto-oncogene p65 Net1; Rho guanine nucleotide exchange factor (GEF) 8; Rho guanine nucleotide exchange factor 8; small GTP-binding protein regulator
Gene Aliases: ARHGEF8; NET1; NET1A
UniProt ID: (Human) Q7Z628
Entrez Gene ID: (Human) 10276
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.