Invitrogen
Manufacturer Code:PA538539
Catalog # PIPA538539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human SDCG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-1; Nuclear export mediator factor NEMF; Nuclear export mediator factor NEMF Antigen NY-CO-1 Serologically defined colon cancer antigen 1 NEMF SDCCAG1; Serologically defined colon cancer antigen 1; Serologically defined colon cancer antigen 1 homolog
Gene Aliases: 1500011I12Rik; 4933405E14Rik; NEMF; NY-CO-1; SDCCAG1
UniProt ID: (Human) O60524, (Mouse) Q8CCP0
Entrez Gene ID: (Human) 9147, (Rat) 100322884, (Mouse) 66244
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.