Novus Biologicals
Manufacturer Code:NBP290179AF405
Catalog # NB070672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 55 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human NELL2 (Q99435 Met1-Leu816). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: nel (chicken)-like 2 NEL-like 2 (chicken) NEL-like protein 2 Nel-related protein 2 NRP2neural epidermal growth factor-like 2 protein kinase C-binding protein NELL2; NEL-like protein 2; NEL-related protein 2; neural epidermal growth factor-like 2; Protein kinase C-binding protein NELL2
Gene Aliases: NELL2; NRP2
UniProt ID: (Human) Q99435
Entrez Gene ID: (Human) 4753
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.