Invitrogen
Manufacturer Code:PA542910
Catalog # PIPA542910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NELL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MEL91 protein; NEL-like 2; NEL-like 2 (chicken) NRP2; nel-like 2 homolog; NEL-like protein 2; NEL-related protein 2; neural epidermal growth factor-like 2; Protein kinase C-binding protein NELL2
Gene Aliases: A330108N19Rik; Mel91; NELL2; NRP2; R75516
UniProt ID: (Bovine) A6QR11, (Human) Q99435, (Mouse) Q80UM5
Entrez Gene ID: (Bovine) 524622, (Pig) 100525817, (Horse) 100054944, (Human) 4753, (Dog) 477636, (Rabbit) 103348937, (Mouse) 54003, (Rat) 81734, (Guinea pig) 100729197
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.