R&D Systems
Manufacturer Code:MAB54871SP
Catalog # MAB54871SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 756435 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human NELL1 Arg17-Asn810 Accession # Q92832 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ45906 IDH3GL nel (chicken)-like 1 NEL-like 1 (chicken) NEL-like protein 1 Nel-related protein 1 neural epidermal growth factor-like 1 NRP1 protein kinase C-binding protein NELL1; NEL-like protein 1; Nel-related protein 1; neural epidermal growth factor-like 1; Protein kinase C-binding protein NELL1
Gene Aliases: IDH3GL; NELL1; NRP1
UniProt ID: (Human) Q92832
Entrez Gene ID: (Human) 4745
Molecular Function: annexin calcium-binding protein calmodulin defense/immunity protein intracellular calcium-sensing protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.