Invitrogen
Manufacturer Code:PA572257
Catalog # PIPA572257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 696-728 amino acids from the C-terminal region of human NELL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IDH3GL NRP1 NEL-like protein 1 nel-related protein 1 neural epidermal growth factor-like 1 protein kinase C-binding protein NELL1; NEL-like protein 1; Nel-related protein 1; neural epidermal growth factor-like 1; Protein kinase C-binding protein NELL1
Gene Aliases: B230343H07Rik; IDH3GL; l7R6; NELL1; NRP1
UniProt ID: (Human) Q92832, (Mouse) Q2VWQ2
Entrez Gene ID: (Human) 4745, (Mouse) 338352
Molecular Function: annexin calcium-binding protein calmodulin defense/immunity protein intracellular calcium-sensing protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.