Novus Biologicals
Manufacturer Code:NBP213651
Catalog # NBP213651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: CHCEKTCQVSGLLYRDQDSWVDGDHCRNCTCKSGAVECRRMSCPPLNCSP DSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITE MCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGENSECK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ45906 IDH3GL nel (chicken)-like 1 NEL-like 1 (chicken) NEL-like protein 1 Nel-related protein 1 neural epidermal growth factor-like 1 NRP1 protein kinase C-binding protein NELL1; NEL-like protein 1; Nel-related protein 1; neural epidermal growth factor-like 1; Protein kinase C-binding protein NELL1
Gene Aliases: IDH3GL; NELL1; NRP1
UniProt ID: (Human) Q92832
Entrez Gene ID: (Human) 4745
Molecular Function: annexin calcium-binding protein calmodulin defense/immunity protein intracellular calcium-sensing protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.