Proteintech
Manufacturer Code:107051AP150UL
Catalog # 50-172-6527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NELFE Fusion Protein Ag1186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D6S45 Negative elongation factor E NELF E NELFE RD RD RNA binding protein RDBP RDP RNA binding protein RD; major histocompatibility complex gene RD; Negative elongation factor E; negative elongation factor polypeptide E; NELF-E; nuclear protein; RD RNA binding protein; RD RNA-binding protein; RNA-binding protein RD
Gene Aliases: D17H6S45; D6S45; NELF-E; NELFE; RD; RDBP; RDP
UniProt ID: (Human) P18615, (Mouse) P19426
Entrez Gene ID: (Human) 7936, (Mouse) 27632, (Rat) 294258
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.