Invitrogen
Manufacturer Code:PA529422
Catalog # PIPA529422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 214 of Human NEK8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JCK NEK12A NPHP9 NIMA-family kinase NEK8 NIMA-related kinase 12a nephrocystin 9 never in mitosis A-related kinase 8 nimA-related protein kinase 8 nima-related protein kinase 12a serine/threonine-protein kinase Nek8 serine/thrionine-protein kinase NEK8; nephrocystin 9; Never in mitosis A-related kinase 8; NIMA (never in mitosis gene a)- related kinase 8; NIMA-family kinase NEK8; NIMA-related kinase 12a; NIMA-related kinase 8; Nima-related protein kinase 12a; NimA-related protein kinase 8; Serine/threonine-protein kinase Nek8; serine/thrionine-protein kinase NEK8
Gene Aliases: JCK; NEK12A; NEK8; NPHP9; RHPD2
UniProt ID: (Human) Q86SG6
Entrez Gene ID: (Human) 284086
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.