Invitrogen
Manufacturer Code:PA5102327
Catalog # PIPA5102327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NEK8 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4632401F23Rik b2b1449Clo JCK juvenile cystic kidney NEK 8 NEK12A Nek8 nephrocystin 9 never in mitosis A-related kinase 8 NIMA (never in mitosis gene a)- related kinase 8 NIMA (never in mitosis gene a)-related expressed kinase 8 NIMA related kinase 8 NIMA-family kinase NEK8 NIMA-related expressed kinase 8 NIMA-related kinase 12a NIMA-related kinase 8 nima-related protein kinase 12a NimA-related protein kinase 8 NPHP9 RHPD2 Serine/threonine-protein kinase Nek8 serine/thrionine-protein kinase NEK8; juvenile cystic kidney; nephrocystin 9; Never in mitosis A-related kinase 8; NIMA (never in mitosis gene a)- related kinase 8; NIMA (never in mitosis gene a)-related expressed kinase 8; NIMA-family kinase NEK8; NIMA-related expressed kinase 8; NIMA-related kinase 12a; NIMA-related kinase 8; Nima-related protein kinase 12a; NimA-related protein kinase 8; Serine/threonine-protein kinase Nek8; serine/thrionine-protein kinase NEK8
Gene Aliases: 4632401F23Rik; b2b1449Clo; JCK; NEK12A; NEK8; NPHP9; RHPD2
UniProt ID: (Human) Q86SG6, (Mouse) Q91ZR4, (Rat) D3ZGQ5
Entrez Gene ID: (Human) 284086, (Mouse) 140859, (Rat) 287473
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.