Invitrogen
Manufacturer Code:MA538018
Catalog # PIMA538018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2342 |
Immunogen | A synthesized peptide derived from human NEK7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810460C19Rik AU020186 NEK7 never in mitosis A-related kinase 7 NIMA (never in mitosis gene a)-related expressed kinase 7 NIMA (never in mitosis gene a)-related kinase 7 NIMA related kinase 7 NIMA-related expressed kinase 7 NIMA-related kinase 7 nimA-related protein kinase 7 Serine/threonine-protein kinase Nek7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; NimA-related protein kinase 7; Serine/threonine-protein kinase Nek7
Gene Aliases: 2810460C19Rik; AU020186; NEK7
UniProt ID: (Human) Q8TDX7, (Rat) D3ZBE5, (Mouse) Q9ES74
Entrez Gene ID: (Human) 140609, (Rat) 360850, (Mouse) 59125
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.