Novus Biologicals
Manufacturer Code:NBP168866
Catalog # NBP168866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CINPDPEKRPDVT corresponding to C-Terminus according to NP_598001.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11.1 Never in mitosis A-related kinase 7 NIMA (never in mitosis gene a)-related kinase 7 NimA-related protein kinase 7 serine/threonine-protein kinase Nek7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA-related kinase 7; NimA-related protein kinase 7; Serine/threonine-protein kinase Nek7
Gene Aliases: NEK7
UniProt ID: (Human) Q8TDX7
Entrez Gene ID: (Human) 140609
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.