Invitrogen
Manufacturer Code:PA5101860
Catalog # PIPA5101860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NEK5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6530411J22 BB049667 Nek5 never in mitosis A-related kinase 5 NIMA (never in mitosis gene a)-related expressed kinase 5 NIMA (never in mitosis gene a)-related kinase 5 NIMA related kinase 5 NIMA-related kinase 5 nimA-related protein kinase 5 Serine/threonine-protein kinase Nek5; Never in mitosis A-related kinase 5; NIMA (never in mitosis gene a)-related kinase 5; NIMA-related kinase 5; NimA-related protein kinase 5; Serine/threonine-protein kinase Nek5
Gene Aliases: 6530411J22; BB049667; NEK5
UniProt ID: (Human) Q6P3R8, (Mouse) Q7TSC3
Entrez Gene ID: (Human) 341676, (Mouse) 330721
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.