Invitrogen
Manufacturer Code:PA5111471
Catalog # PIPA5111471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NEK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nek4 Never in mitosis A-related kinase 4 NimA-related protein kinase 4 NRK2 pp12301 Serine/threonine-protein kinase 2 Serine/threonine-protein kinase Nek4 Serine/threonine-protein kinase NRK2 STK2; Never in mitosis A-related kinase 4; NIMA (never in mitosis gene a)-related kinase 4; NIMA-related kinase 4; NimA-related protein kinase 4; serine/threonine kinase 2; serine/threonine protein kinase-2; Serine/threonine-protein kinase 2; Serine/threonine-protein kinase Nek4; serine/threonine-protein kinase Nek4 variant 2; Serine/threonine-protein kinase NRK2
Gene Aliases: NEK4; NRK2; pp12301; STK2
UniProt ID: (Human) P51957
Entrez Gene ID: (Human) 6787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.