Invitrogen
Manufacturer Code:PA5104126
Catalog # PIPA5104126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NEK4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nek4 never in mitosis A-related kinase 4 NIMA (never in mitosis gene a)-related expressed kinase 4 NIMA (never in mitosis gene a)-related kinase 4 NIMA related kinase 4 NIMA-related kinase 4 NimA-related protein kinase 4 NRK2 pp12301 serine/threonine kinase 2 serine/threonine protein kinase-2 serine/threonine-protein kinase 2 Serine/threonine-protein kinase Nek4 serine/threonine-protein kinase Nek4 variant 2 Serine/threonine-protein kinase NRK2 STK2; Never in mitosis A-related kinase 4; NIMA (never in mitosis gene a)-related expressed kinase 4; NIMA (never in mitosis gene a)-related kinase 4; NIMA-related kinase 4; NimA-related protein kinase 4; serine/threonine kinase 2; serine/threonine protein kinase-2; Serine/threonine-protein kinase 2; Serine/threonine-protein kinase Nek4; serine/threonine-protein kinase Nek4 variant 2; Serine/threonine-protein kinase NRK2
Gene Aliases: NEK4; NRK2; pp12301; STK2
UniProt ID: (Human) P51957, (Mouse) Q9Z1J2
Entrez Gene ID: (Human) 6787, (Mouse) 23955, (Rat) 306252
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.