Novus Biologicals
Manufacturer Code:NBP298357IR
Catalog # NB082129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant NEK3 (ABK42288.1 Met1-Ala509). |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 glycogen synthase A kinase HSPK 36 HSPK36 hydroxyalkyl-protein kinase MGC29949 Never in mitosis A-related kinase 3 NIMA (never in mitosis gene a)-related kinase 3 NimA-related protein kinase 3 phosphorylase B kinase kinase serine/threonine-protein kinase Nek3; glycogen synthase A kinase; HSPK 36; hydroxyalkyl-protein kinase; Never in mitosis A-related kinase 3; NIMA (never in mitosis gene a)-related kinase 3; NIMA-related kinase 3; NimA-related protein kinase 3; phosphorylase B kinase kinase; Serine/threonine-protein kinase Nek3
Gene Aliases: HSPK36; NEK3
UniProt ID: (Human) P51956
Entrez Gene ID: (Human) 4752
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.