Invitrogen
Manufacturer Code:PA592416
Catalog # PIPA592416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-135 of human NEK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPK 21; Nek2 kinase; Never in mitosis A-related kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NIMA - related expressed kinase 2; NimA-like protein kinase 1; NIMA-related expressed kinase 2; NIMA-related kinase 2; NimA-related protein kinase 2; protein phosphatase 1, regulatory subunit 111; Serine/threonine-protein kinase Nek2
Gene Aliases: AA617254; C77054; HsPK21; NEK2; NEK2A; NLK1; PPP1R111; RP67
UniProt ID: (Human) P51955, (Mouse) O35942
Entrez Gene ID: (Human) 4751, (Mouse) 18005
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.