OriGene
Manufacturer Code:CF501608
Catalog # 50-166-0606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E9 |
Immunogen | Full length human recombit protein of human NEK11 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA related kinase 11; NimA-related protein kinase 11; Serine/threonine-protein kinase Nek11
Gene Aliases: NEK11
UniProt ID: (Human) Q8NG66
Entrez Gene ID: (Human) 79858
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.