Novus Biologicals
Manufacturer Code:NBP272935
Catalog # NBP272935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D9 |
Immunogen | Full length human recombinant protein of human NEK11(NP_079076) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.11 EC 2.7.11.1 FLJ23495 Never in mitosis A-related kinase 11 NIMA (never in mitosis gene a)- related kinase 11 NimA-related protein kinase 11 serine/threonine-protein kinase Nek11; Never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NimA-related protein kinase 11; Serine/threonine-protein kinase Nek11
Gene Aliases: NEK11
UniProt ID: (Human) Q8NG66
Entrez Gene ID: (Human) 79858
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.