Invitrogen
Manufacturer Code:PA515334
Catalog # PIPA515334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 480-510 amino acids from the C-terminal region of human NEK11L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: nek11; Never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA-related kinase 11; NIMA-related kinase 11L; NimA-related protein kinase 11; Serine/threonine-protein kinase Nek11
Gene Aliases: 4932416N14Rik; NEK11
UniProt ID: (Human) Q8NG66, (Mouse) Q8C0Q4
Entrez Gene ID: (Human) 79858, (Mouse) 208583
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.