Invitrogen
Manufacturer Code:PA558601
Catalog # PIPA558601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NEK10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Never in mitosis A-related kinase 10; NIMA (never in mitosis gene a)- related kinase 10; NIMA-related kinase 10; NimA-related protein kinase 10; Serine/threonine-protein kinase Nek10; Serine/threonine-protein kinase Nek10 Never in mitosis A-related kinase 10 NimA-related protein kinase 10 NEK10
Gene Aliases: NEK10
UniProt ID: (Human) Q6ZWH5
Entrez Gene ID: (Human) 152110
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.