Novus Biologicals
Manufacturer Code:NBP182884
Catalog # NBP182884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:NLKAQEDEKGKQNLSDTFEINVHEDAKEHEKEKSVSSDRKKWEAGGQLVIPLDELTLDTSFSTTERHTVGEVIKLGPNGSPRRAWGKSPTDSVLKILGEAELQLQTELLENTTIRSEISPEGEKYKPLITGEKKVQCISHEINPSA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686D06121 DKFZp686K12169 EC 2.7.11 EC 2.7.11.1 KIAA1901MGC138800 Never in mitosis A-related kinase 1 NIMA (never in mitosis gene a)-related kinase 1 NimA-related protein kinase 1 NY-REN-55 protein-serine/threonine kinase Renal carcinoma antigen NY-REN-55 serine/threonine-protein kinase Nek1 SRPS2; Never in mitosis A-related kinase 1; NIMA (never in mitosis gene a)-related kinase 1; NIMA-related kinase 1; NimA-related protein kinase 1; protein-serine/threonine kinase; Renal carcinoma antigen NY-REN-55; Serine/threonine-protein kinase Nek1
Gene Aliases: KIAA1901; NEK1; NY-REN-55; SRPS2; SRPS2A; SRTD6
UniProt ID: (Human) Q96PY6
Entrez Gene ID: (Human) 4750
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.