Abnova
Manufacturer Code:PAB3282
Catalog # 89-130-927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human NEK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NEK1.
For Research Use Only
Protein Aliases: DKFZp686D06121; DKFZp686K12169; KIAA1901; MGC138800; Never in mitosis A-related kinase 1; NIMA (never in mitosis gene a)-related kinase 1; NIMA-related kinase 1; NimA-related protein kinase 1; NY-REN-55; protein-serine/threonine kinase; Renal carcinoma antigen NY-REN-55; Serine/threonine-protein kinase Nek1
Gene Aliases: KIAA1901; NEK1; NY-REN-55; SRPS2; SRPS2A; SRTD6
UniProt ID: (Human) Q96PY6
Entrez Gene ID: (Human) 4750
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.