Invitrogen
Manufacturer Code:PA5101580
Catalog # PIPA5101580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NEIL3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI449477 BC034753 C85903 DNA glycosylase FPG2 DNA glycosylase hFPG2 DNA glycosylase/AP lyase Neil3 Endonuclease 8-like 3 endonuclease VIII-like 3 FGP2 FLJ10858 FPG2 hFPG2 hNEI3 nei endonuclease VIII-like 3 nei like 3 (E. coli) nei like DNA glycosylase 3 NEI3 Neil3 nei-like DNA glycosylase 3 nei-like protein 3 ZGRF3 zinc finger GRF-type containing 3; DNA glycosylase FPG2; DNA glycosylase hFPG2; DNA glycosylase/AP lyase Neil3; Endonuclease 8-like 3; Endonuclease VIII-like 3; nei endonuclease VIII-like 3; nei-like DNA glycosylase 3; Nei-like protein 3; zinc finger, GRF-type containing 3
Gene Aliases: AI449477; BC034753; C85903; FGP2; FPG2; hFPG2; hNEI3; NEI3; NEIL3; ZGRF3
UniProt ID: (Human) Q8TAT5, (Mouse) Q8K203
Entrez Gene ID: (Human) 55247, (Mouse) 234258
Molecular Function: DNA binding protein DNA glycosylase damaged DNA-binding protein endodeoxyribonuclease esterase glycosidase hydrolase lyase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.