Abnova
Manufacturer Code:PAB1054
Catalog # 89-114-857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 164-177 of human NEI3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NEIL3.
For Research Use Only
Protein Aliases: DNA glycosylase FPG2; DNA glycosylase hFPG2; DNA glycosylase/AP lyase Neil3; Endonuclease 8-like 3; Endonuclease VIII-like 3; FGP2 FLJ10858 FPG2 NEI3 hFPG2 hNEI3; nei endonuclease VIII-like 3; nei-like DNA glycosylase 3; Nei-like protein 3; zinc finger, GRF-type containing 3
Gene Aliases: FGP2; FPG2; hFPG2; hNEI3; NEI3; NEIL3; ZGRF3
UniProt ID: (Human) Q8TAT5
Entrez Gene ID: (Human) 55247
Molecular Function: DNA binding protein DNA glycosylase damaged DNA-binding protein endodeoxyribonuclease esterase glycosidase hydrolase lyase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.