Proteintech
Manufacturer Code:116211AP150UL
Catalog # 50-172-7317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NEIL3 Fusion Protein Ag2200 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA glycosylase FPG2; DNA glycosylase FPG2 DNA glycosylase/AP lyase Neil3 Endonuclease 8 like 3 Endonuclease VIII like 3 FGP2 FPG2 hFPG2 hNEI3 Nei like protein 3 NEI3 NEIL3; DNA glycosylase hFPG2; DNA glycosylase/AP lyase Neil3; Endonuclease 8-like 3; Endonuclease VIII-like 3; nei endonuclease VIII-like 3; nei-like DNA glycosylase 3; Nei-like protein 3; zinc finger, GRF-type containing 3
Gene Aliases: AI449477; BC034753; C85903; FGP2; FPG2; hFPG2; hNEI3; NEI3; NEIL3; ZGRF3
UniProt ID: (Human) Q8TAT5, (Mouse) Q8K203
Entrez Gene ID: (Human) 55247, (Mouse) 234258
Molecular Function:
DNA binding protein
DNA glycosylase
damaged DNA-binding protein
endodeoxyribonuclease
esterase
glycosidase
hydrolase
lyase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.