Novus Biologicals
Manufacturer Code:NBP272932H
Catalog # NBP272932H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 186-390 of human NEIL1 (NP_078884) produced in E.coli. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1 EC 3.2.2.- EC 4.2.99.18 endonuclease 8-like 1 endonuclease VIII Endonuclease VIII-like 1 FLJ22402 FPG1DNA glycosylase/AP lyase Neil1 hFPG1 NEH1 nei endonuclease VIII-like 1 (E. coli) Nei homolog 1 NEI1 Nei-like protein 1; Endonuclease 8-like 1; endonuclease VIII; Endonuclease VIII-like 1; FPG1; NEH1; nei endonuclease VIII-like 1; Nei homolog 1; nei-like DNA glycosylase 1; Nei-like protein 1
Gene Aliases: FPG1; hFPG1; NEI1; NEIL1
UniProt ID: (Human) Q96FI4
Entrez Gene ID: (Human) 79661
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.