Abnova
Manufacturer Code:H00079661K
Catalog # 89-204-158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NEIL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NEIL1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; Endonuclease 8-like 1; endonuclease VIII; Endonuclease VIII-like 1; FLJ22402 FPG1 NEI1 hFPG1; FPG1; NEH1; nei endonuclease VIII-like 1; Nei homolog 1; nei-like DNA glycosylase 1; Nei-like protein 1
Gene Aliases: FPG1; hFPG1; NEI1; NEIL1
UniProt ID: (Human) Q96FI4
Entrez Gene ID: (Human) 79661
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.