Invitrogen
Manufacturer Code:PA585229
Catalog # PIPA585229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human NEGR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMML2433 antibody IGLON4 antibody KILON antibody MGC46680 antibody Ntra antibody NEGR1 antibody neurotractin antibody IgLON family member 4 antibody a kindred of IgLON antibody neuronal growth regulator 1 antibody; Kilon; Kindred of IgLON; Neuronal growth regulator 1; Neurotractin
Gene Aliases: 5330422G01Rik; Kiaa3001; KILON; Negr1; Ntra
UniProt ID: (Mouse) Q80Z24
Entrez Gene ID: (Mouse) 320840
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.