OriGene
Manufacturer Code:CF805823
Catalog # 50-166-7580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 413-773 of human NEFM produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 160 kDa neurofilament protein; NEF3 NF-M NFM; Neurofilament 3; Neurofilament medium polypeptide; Neurofilament triplet M protein; neurofilament, medium polypeptide 150kDa; neurofilament-3 (150 kD medium); NF-M
Gene Aliases: NEF3; NEFM; NF-M; NFM
UniProt ID: (Human) P07197
Entrez Gene ID: (Human) 4741
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.