Invitrogen
Manufacturer Code:PA116758
Catalog # APPA6758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | The C-terminal extension of rat NF-M (KE segment) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: 160 kDa neurofilament protein; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; Neurofilament medium polypeptide; neurofilament protein M; Neurofilament protein, middle polypeptide; Neurofilament triplet M protein; neurofilament, medium polypeptide 150kDa; neurofilament-3 (150 kD medium); neurofilament-M; NF-M; NF-M c-terminus; NF-M protein; NFM NF-M NMC160 kD neurofilament protein neurofilament medium polypeptide neurofilament 3 NEF3
Gene Aliases: NEF3; NEFM; NF-M; NF160; NF165; NFM
UniProt ID: (Human) P07197, (Chicken) P16053, (Mouse) P08553, (Rat) P12839
Entrez Gene ID: (Human) 4741, (Chicken) 396206, (Mouse) 18040, (Rat) 24588
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.