Invitrogen
Manufacturer Code:MA515097
Catalog # PIMA515097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F.852.7 |
Immunogen | Purified and enzymatically dephosphorylated pig neurofilament light chain |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 68 kDa neurofilament protein; 68 kDa neurofilament protein OTTHUMP00000196614 light molecular weight neurofilament protein neurofilament light polypeptide neurofilament protein light chain neurofilament subunit NF-L neurofilament triplet L protein neurofilament light polypeptide 68kDa neurofilament-light; light molecular weight neurofilament protein; Neurofilament light polypeptide; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet L protein; neurofilament, light polypeptide 68kDa; NF-L; protein phosphatase 1, regulatory subunit 110
Gene Aliases: AI847934; CMT1F; CMT2E; NEFL; NF-L; NF68; NFL; PPP1R110
UniProt ID: (Human) P07196, (Mouse) P08551, (Rat) P19527
Entrez Gene ID: (Human) 4747, (Mouse) 18039, (Rat) 83613
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.