Abnova
Manufacturer Code:H00004747D01
Catalog # 89-002-905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NEFL (NP_006149.2 1 a.a. ~ 543 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human NEFL protein.
For Research Use Only
Protein Aliases: 68 kDa neurofilament protein; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; Neurofilament light polypeptide; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet L protein; neurofilament, light polypeptide 68kDa; NF-L; NF68; NFL; protein phosphatase 1, regulatory subunit 110
Gene Aliases: CMT1F; CMT2E; NEFL; NF-L; NF68; NFL; PPP1R110
UniProt ID: (Human) P07196
Entrez Gene ID: (Human) 4747
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.