Novus Biologicals
Manufacturer Code:NB11059137
Catalog # NB11059137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ND23 |
Immunogen | Synthetic peptide corresponding to amino acids 20-32 of human Nedd8 conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ43224 MGC104393 MGC125896 MGC125897 NEDD-8 Neddylin Neural precursor cell expressed developmentally down-regulated protein 8 neural precursor cell expressed developmentally down-regulated 8 Ubiquitin-like protein Nedd8; NEDD-8; NEDD8; Neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; Ubiquitin-like protein Nedd8
Gene Aliases: NEDD-8; NEDD8
UniProt ID: (Human) Q15843
Entrez Gene ID: (Human) 4738
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.