Novus Biologicals
Manufacturer Code:NBP237523
Catalog # NBP237523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B8 |
Immunogen | Purified recombinant fragment of human NEDD8 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ43224 MGC104393 MGC125896 MGC125897 NEDD-8 Neddylin Neural precursor cell expressed developmentally down-regulated protein 8 neural precursor cell expressed developmentally down-regulated 8 Ubiquitin-like protein Nedd8; NEDD-8; NEDD8; Neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; Ubiquitin-like protein Nedd8
Gene Aliases: NEDD-8; NEDD8
UniProt ID: (Human) Q15843
Entrez Gene ID: (Human) 4738
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.