R&D Systems
Manufacturer Code:MAB4936SP
Catalog # MAB4936SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 584606 |
Immunogen | <EM>E. coli</EM>-derived recombinant human NEDD8 Leu2-Gly77 Accession # Q15843 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ43224 MGC104393 MGC125896 MGC125897 NEDD-8 Neddylin Neural precursor cell expressed developmentally down-regulated protein 8 neural precursor cell expressed developmentally down-regulated 8 Rub1 Ubiquitin-like protein Nedd8; NEDD-8; NEDD8; Neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; Ubiquitin-like protein Nedd8
Gene Aliases: NEDD-8; NEDD8
UniProt ID: (Human) Q15843
Entrez Gene ID: (Human) 4738
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.